Heptyl-p-toluenesulphonate 98%

Synonyme: Heptyl 4-methylbenzenesulphonate,p-Toluenesulphonic acid Heptyl Ester

BD15639-5G BD15639-25G BD15639-100G BD15639-10G BD15639-1G
BLDPBD15639-5GEA 21 EUR
BLDPBD15639-5G BLDPBD15639-25G BLDPBD15639-100G BLDPBD15639-10G BLDPBD15639-1G
Heptyl-p-toluenesulphonate 98%
Heptyl-p-toluenesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₂₂O₃S
Molecular Weight: 270,39 g/mol
Lagertemperatur: Ambient
MDL: MFCD00059442
CAS-Nummer: 24767-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR