Valethamate bromide 98%

BLDPBD156285-5GEA 295 EUR
BLDPBD156285-5G BLDPBD156285-1G BLDPBD156285-250MG
Valethamate bromide 98%
Valethamate bromide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₉H₃₂BrNO₂
Molecular Weight: 386,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031617
CAS-Nummer: 90-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR