Valeramid 98%

BLDPBD156275-1GEA 7 EUR
BLDPBD156275-1G BLDPBD156275-5G BLDPBD156275-100G BLDPBD156275-25G
Valeramid 98%
Valeramid

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₁₀O₂
Molecular Weight: 102,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00041895
CAS-Nummer: 626-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR