Tyrphostin B46 98%

BLDPBD156127-50MGEA 119 EUR
BLDPBD156127-50MG
Tyrphostin B46 98%
Tyrphostin B46

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₉H₁₈N₂O₃
Molecular Weight: 322,36 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00209865
CAS-Nummer: 133550-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR