Tyrphostin AG-1296 99+%

Synonyme: 6,7-Dimethoxy-2-phenylquinoxaline

BD156123-250MG BD156123-25MG BD156123-100MG BD156123-1G
BLDPBD156123-250MGEA 328 EUR
BLDPBD156123-250MG BLDPBD156123-25MG BLDPBD156123-100MG BLDPBD156123-1G
Tyrphostin AG-1296 99+%
Tyrphostin AG-1296

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₆H₁₄N₂O₂
Molecular Weight: 266,3 g/mol
Lagertemperatur: Ambient
MDL: MFCD00270913
CAS-Nummer: 146535-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR