Tyrphostin AG-126 98%

Synonyme: 2-(3-Hydroxy-4-nitrobenzylidene)malononitrile

BD156121-10MG BD156121-5MG
BLDPBD156121-10MGEA 132 EUR
BLDPBD156121-10MG BLDPBD156121-5MG
Tyrphostin AG-126 98%
Tyrphostin AG-126

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₀H₅N₃O₃
Molecular Weight: 215,168 g/mol
Lagertemperatur: Freezer
MDL: MFCD00236448
CAS-Nummer: 118409-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR