1-Bromo-8-phenyloctane 95%

Synonyme: (8-Bromooct-1-yl)benzene

BD15610-5G BD15610-1G
BLDPBD15610-5GEA 215 EUR
BLDPBD15610-5G BLDPBD15610-1G
1-Bromo-8-phenyloctane 95%
1-Bromo-8-phenyloctane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₄H₂₁Br
Molecular Weight: 269,221 g/mol
Lagertemperatur: Ambient
MDL: MFCD01075178
CAS-Nummer: 54646-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR