Tryptanthrin 97%

Synonyme: Indolo[2,1-b]quinazoline-6,12-dione

BD156048-250MG BD156048-10G BD156048-1G BD156048-5G BD156048-100MG BD156048-25G
BLDPBD156048-250MGEA 39 EUR
BLDPBD156048-250MG BLDPBD156048-10G BLDPBD156048-1G BLDPBD156048-5G BLDPBD156048-100MG BLDPBD156048-25G
Tryptanthrin 97%
Tryptanthrin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₈N₂O₂
Molecular Weight: 248,2362 g/mol
Lagertemperatur: Ambient
MDL: MFCD00012073
CAS-Nummer: 13220-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR