tri-Natriumphosphat Dodecahydrat 99%

Synonyme: Natriumphosphat tribasisch Dodecahydrat

BLDPBD155947-100GEA 8 EUR
BLDPBD155947-100G BLDPBD155947-1000G BLDPBD155947-500G
tri-Natriumphosphat Dodecahydrat 99%
tri-Natriumphosphat Dodecahydrat

Storage condition: Sealed in dry, Room Temperature

Formel: Na₃PO₄·12H₂O
Molecular Weight: 380,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149198
CAS-Nummer: 10101-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR