Tris[2-(dimethylamino)ethyl]amine 99%

Synonyme: Me6TREN

BD155802-250MG BD155802-5G BD155802-25G BD155802-1G
BLDPBD155802-250MGEA 8 EUR
BLDPBD155802-250MG BLDPBD155802-5G BLDPBD155802-25G BLDPBD155802-1G
Tris[2-(dimethylamino)ethyl]amine 99%
Tris[2-(dimethylamino)ethyl]amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₃₀N₄
Molecular Weight: 230,40 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015607
CAS-Nummer: 33527-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR