Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate 98%

Synonyme: Tris(2,2'-bipyridyl)dichlororuthenium(II),Dichlorotris(2,2'-bipyridyl)ruthenium(II),Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate,Tris(2,2-bipyridyl)ruthenium(II) chloride hexahydrate

BD155782-1G BD155782-10G BD155782-5G BD155782-25G
BLDPBD155782-1GEA 27 EUR
BLDPBD155782-1G BLDPBD155782-10G BLDPBD155782-5G BLDPBD155782-25G
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₃₀H₃₆Cl₂N₆O₆Ru
Molecular Weight: 748,62 g/mol
Lagertemperatur: Ambient
MDL: MFCD00149670
CAS-Nummer: 50525-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR