Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite 98%

Synonyme: Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphite,Phosphorous acid tris(1,1,1,3,3,3-hexafluoro-2-propyl) ester

BD155770-5G BD155770-1G BD155770-250MG
BLDPBD155770-5GEA 237 EUR
BLDPBD155770-5G BLDPBD155770-1G BLDPBD155770-250MG
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite 98%
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₃F₁₈O₃P
Molecular Weight: 532,07 g/mol
Lagertemperatur: Ambient
MDL: MFCD00192561
CAS-Nummer: 66470-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR