1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol 98%

BD155752-100G BD155752-500G BD155752-1000G BD155752-25G
BLDPBD155752-100GEA 25 EUR
BLDPBD155752-100G BLDPBD155752-500G BLDPBD155752-1000G BLDPBD155752-25G
1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol 98%
1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₂O₄
Molecular Weight: 206,27 g/mol
Lagertemperatur: Ambient
MDL: MFCD00059142
CAS-Nummer: 20324-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR