Tripropylorthoformiat 95%

Synonyme: 1,1',1''-[Methylidyntris(oxy)]trispropan,Tripropoxymethan,Orthoameisensäuretripropylester

BD155749-25G BD155749-1G BD155749-100G BD155749-5G
BLDPBD155749-25GEA 70 EUR
BLDPBD155749-25G BLDPBD155749-1G BLDPBD155749-100G BLDPBD155749-5G
Tripropylorthoformiat 95%
Tripropylorthoformiat

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₂₂O₃
Molecular Weight: 190,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00015214
CAS-Nummer: 621-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR