Triphenylmethan 98%

Synonyme: Tritan

BLDPBD155706-25GEA 7 EUR
BLDPBD155706-25G BLDPBD155706-5G BLDPBD155706-500G BLDPBD155706-1000G BLDPBD155706-100G
Triphenylmethan 98%
Triphenylmethan

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₉H₁₆
Molecular Weight: 244,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004763
CAS-Nummer: 519-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR