Tricaprylin 98%

Synonyme: Propane-1,2,3-triyl trioctanoate,1,2,3-Tri-n-octanoylglycerol,Tri-n-octanoin,Glycerol Tri-n-octanoate

BD155646-25G BD155646-100G BD155646-5G BD155646-500G
BLDPBD155646-25GEA 31 EUR
BLDPBD155646-25G BLDPBD155646-100G BLDPBD155646-5G BLDPBD155646-500G
Tricaprylin 98%
Tricaprylin

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₇H₅₀O₆
Molecular Weight: 470,69 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00036236
CAS-Nummer: 538-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR