Tri-O-benzyl-D-glucal 97%

BLDPBD155644-10GEA 320 EUR
BLDPBD155644-10G BLDPBD155644-25G BLDPBD155644-1G BLDPBD155644-5G BLDPBD155644-250MG
Tri-O-benzyl-D-glucal 97%
Tri-O-benzyl-D-glucal

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₇H₂₈O₄
Molecular Weight: 416,52 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00061640
CAS-Nummer: 55628-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR