Trimethylsilyl 2-((trimethylsilyl)oxy)acetate 97%

BLDPBD155546-25GEA 50 EUR
BLDPBD155546-25G BLDPBD155546-5G
Trimethylsilyl 2-((trimethylsilyl)oxy)acetate 97%
Trimethylsilyl 2-((trimethylsilyl)oxy)acetate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₂₀O₃Si₂
Molecular Weight: 220,41 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009840
CAS-Nummer: 33581-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR