Trimethylsilylacetat (Acetoxytrimethylsilan) 98%

BLDPBD155527-5GEA 6 EUR
BLDPBD155527-5G BLDPBD155527-25G
Trimethylsilylacetat (Acetoxytrimethylsilan) 98%
Trimethylsilylacetat (Acetoxytrimethylsilan)

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₁₂O₂Si
Molecular Weight: 132,24 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00008695
CAS-Nummer: 2754-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR