Trimellitsäure 98%

Synonyme: 1,2,4-Benzoltricarbonsäure,Benzol-1,2,4-tricarbonsäure

BD155393-5G BD155393-500G BD155393-100G BD155393-25G
BLDPBD155393-5GEA 7 EUR
BLDPBD155393-5G BLDPBD155393-500G BLDPBD155393-100G BLDPBD155393-25G
Trimellitsäure 98%
Trimellitsäure

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₆O₆
Molecular Weight: 210,14 g/mol
Lagertemperatur: Ambient
MDL: MFCD00002470
CAS-Nummer: 528-44-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR