6-Brom-2-naphthyl-trifluormethansulfonat 95%

Synonyme: 6-Bromo-2-naphthyl Triflate,Trifluoromethanesulphonic acid 6-Bromo-2-naphthyl Ester,6-Bromonaphthalen-2-yl trifluoromethanesulphonate

BD155301-25G BD155301-250MG BD155301-1G BD155301-5G
BLDPBD155301-25GEA 563 EUR
BLDPBD155301-25G BLDPBD155301-250MG BLDPBD155301-1G BLDPBD155301-5G
6-Brom-2-naphthyl-trifluormethansulfonat 95%
6-Brom-2-naphthyl-trifluormethansulfonat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₆BrF₃O₃S
Molecular Weight: 355,13 g/mol
Lagertemperatur: Ambient
MDL: MFCD03093642
CAS-Nummer: 151600-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR