Triethylene glycol monobutyl ether 98% (durch GC)

Synonyme: Triethyleneglycolmonobutylether,Butyl Triglycol

BD155230-100G BD155230-1000G BD155230-500G
BLDPBD155230-100GEA 7 EUR
BLDPBD155230-100G BLDPBD155230-1000G BLDPBD155230-500G
Triethylene glycol monobutyl ether 98% (durch GC)
Triethylene glycol monobutyl ether

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₂O₄
Molecular Weight: 206,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00020606
CAS-Nummer: 143-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR