Triethylenglycoldimethylether 99% stabilisiert

Synonyme: 2,5,8,11-Tetraoxadodecane

Stabilisierung: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

BD155228-25G
BLDPBD155228-25GEA 6 EUR
BLDPBD155228-25G
Triethylenglycoldimethylether 99% stabilisiert
Triethylenglycoldimethylether

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₈O₄
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008504
CAS-Nummer: 112-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR