Fmoc-3-(3-pyridyl)-L-alanine 97%

BLDPBD1552-25GEA 336 EUR
BLDPBD1552-25G BLDPBD1552-100G BLDPBD1552-5G BLDPBD1552-10G BLDPBD1552-250MG BLDPBD1552-1G
Fmoc-3-(3-pyridyl)-L-alanine 97%
Fmoc-3-(3-pyridyl)-L-alanine

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₃H₂₀N₂O₄
Molecular Weight: 388,423 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 175453-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR