Triethyl-3-phosphonopropionat 95%

BLDPBD155190-100GEA 212 EUR
BLDPBD155190-100G BLDPBD155190-10G BLDPBD155190-5G BLDPBD155190-25G BLDPBD155190-1G
Triethyl-3-phosphonopropionat 95%
Triethyl-3-phosphonopropionat

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₉O₅P
Molecular Weight: 238,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00015167
CAS-Nummer: 3699-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR