Triethyl 1,3,5-triazine-2,4,6-tricarboxylate 98%

BLDPBD155181-5GEA 360 EUR
BLDPBD155181-5G BLDPBD155181-1G BLDPBD155181-250MG
Triethyl 1,3,5-triazine-2,4,6-tricarboxylate 98%
Triethyl 1,3,5-triazine-2,4,6-tricarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₂H₁₅N₃O₆
Molecular Weight: 297,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01419268
CAS-Nummer: 898-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR