Boc-3-bromo-L-phenylalanine 98%

Synonyme: Boc-Phe(3-Br)-OH

BD15507-10G BD15507-1G BD15507-250MG BD15507-5G BD15507-25G BD15507-100G
BLDPBD15507-10GEA 70 EUR
BLDPBD15507-10G BLDPBD15507-1G BLDPBD15507-250MG BLDPBD15507-5G BLDPBD15507-25G BLDPBD15507-100G
Boc-3-bromo-L-phenylalanine 98%
Boc-3-bromo-L-phenylalanine

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₈BrNO₄
Molecular Weight: 344,21 g/mol
Lagertemperatur: Ambient
MDL: MFCD01317710
CAS-Nummer: 82278-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR