Tribenzylphosphin 98%

BLDPBD155017-100MGEA 19 EUR
BLDPBD155017-100MG BLDPBD155017-250MG BLDPBD155017-5G BLDPBD155017-1G
Tribenzylphosphin 98%
Tribenzylphosphin

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂₁H₂₁P
Molecular Weight: 304,37 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00014439
CAS-Nummer: 7650-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR