(3R,4R,5R)-4-(Benzyloxy)-5-((R)-1,2-bis(benzyloxy)ethyl)-2-ethoxytetrahydrofuran-3-ol 97%

BD155012-5G BD155012-1G BD155012-10G
BLDPBD155012-5GEA 232 EUR
BLDPBD155012-5G BLDPBD155012-1G BLDPBD155012-10G
(3R,4R,5R)-4-(Benzyloxy)-5-((R)-1,2-bis(benzyloxy)ethyl)-2-ethoxytetrahydrofuran-3-ol 97%
(3R,4R,5R)-4-(Benzyloxy)-5-((R)-1,2-bis(benzyloxy)ethyl)-2-ethoxytetrahydrofuran-3-ol

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₂₉H₃₄O₆
Molecular Weight: 478,57 g/mol
Lagertemperatur: Freezer
MDL: MFCD00801089
CAS-Nummer: 10310-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR