Tri-(p-tert-butylphenyl)phosphate 95%

BD154968-1G BD154968-5G BD154968-250MG
BLDPBD154968-1GEA 63 EUR
BLDPBD154968-1G BLDPBD154968-5G BLDPBD154968-250MG
Tri-(p-tert-butylphenyl)phosphate 95%
Tri-(p-tert-butylphenyl)phosphate

Storage condition: Sealed in dry, Room Temperature

Formel: C₃₀H₃₉O₄P
Molecular Weight: 494,60 g/mol
Lagertemperatur: Ambient
MDL: MFCD00026302
CAS-Nummer: 78-33-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR