Tri(2-furyl)phosphin 98%

Synonyme: Tri(furan-2-yl)phosphine,Tri-2-furylphosphine

BD154957-250MG BD154957-25G BD154957-100G BD154957-500G BD154957-5G BD154957-1G
BLDPBD154957-250MGEA 7 EUR
BLDPBD154957-250MG BLDPBD154957-25G BLDPBD154957-100G BLDPBD154957-500G BLDPBD154957-5G BLDPBD154957-1G
Tri(2-furyl)phosphin 98%
Tri(2-furyl)phosphine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₉O₃P
Molecular Weight: 232,17 g/mol
Lagertemperatur: Ambient
MDL: MFCD00151857
CAS-Nummer: 5518-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR