(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane) 97%

BLDPBD154805-5GEA 81 EUR
BLDPBD154805-5G BLDPBD154805-1G BLDPBD154805-250MG
(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane) 97%
(trans,trans)-4-Ethoxy-4'-propyl-1,1'-bi(cyclohexane)

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₇H₃₂O
Molecular Weight: 252,44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22421614
CAS-Nummer: 95756-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR