(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid 98%

BLDPBD15480-25GEA 326 EUR
BLDPBD15480-25G BLDPBD15480-1G BLDPBD15480-250MG BLDPBD15480-100MG BLDPBD15480-5G
(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid 98%
(R)-1-((Benzyloxy)carbonyl)piperidine-2-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₄H₁₇NO₄
Molecular Weight: 263,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01863738
CAS-Nummer: 28697-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR