trans-4-Brom-β-nitrostyrol 95%

Synonyme: (E)-1-Bromo-4-(2-nitrovinyl)benzene

BD154790-1G BD154790-250MG BD154790-5G
BLDPBD154790-1GEA 55 EUR
BLDPBD154790-1G BLDPBD154790-250MG BLDPBD154790-5G
trans-4-Brom-β-nitrostyrol 95%
trans-4-Brom-β-nitrostyrol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₆BrNO₂
Molecular Weight: 228,0427 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00191856
CAS-Nummer: 5153-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR