Methyl 1-benzyl-4-oxopiperidine-3-carboxylate 95%

BLDPBD15476-1GEA 46 EUR
BLDPBD15476-1G
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate 95%
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00044754
CAS-Nummer: 57611-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR