trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl 97%

Synonyme: 4-Fluoro-4'-(trans-4-propylcyclohexyl)biphenyl,4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

BD154758-10G BD154758-25G BD154758-100G BD154758-1G BD154758-5G
BLDPBD154758-10GEA 34 EUR
BLDPBD154758-10G BLDPBD154758-25G BLDPBD154758-100G BLDPBD154758-1G BLDPBD154758-5G
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl 97%
trans-4-Fluoro-4'-(4-n-propylcyclohexyl)biphenyl

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₁H₂₅F
Molecular Weight: 296,429 g/mol
Lagertemperatur: Ambient
MDL: MFCD11053438
CAS-Nummer: 87260-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR