trans-3-(Trifluoromethyl)cinnamoyl chloride 98%

Synonyme: (E)-3-(3-(Trifluoromethyl)phenyl)acryloyl chloride

BD154720-1G BD154720-250MG BD154720-5G BD154720-25G
BLDPBD154720-1GEA 39 EUR
BLDPBD154720-1G BLDPBD154720-250MG BLDPBD154720-5G BLDPBD154720-25G
trans-3-(Trifluoromethyl)cinnamoyl chloride 98%
trans-3-(Trifluoromethyl)cinnamoyl chloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₆ClF₃O
Molecular Weight: 234,602 g/mol
Lagertemperatur: Ambient
MDL: MFCD00075514
CAS-Nummer: 64379-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR