trans-2-Dodecenoic acid 98%
|
|
BLDPBD154666-100MGEA
43
EUR
BLDPBD154666-100MG
BLDPBD154666-1G
BLDPBD154666-250MG
BLDPBD154666-5G
trans-2-Dodecenoic acid 98%
trans-2-Dodecenoic acid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₂H₂₂O₂ Molecular Weight: 198,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045906 CAS-Nummer: 32466-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...