Monomethyl-trans-1,4-cyclohexanedicarboxylate 98%

BLDPBD154575-25GEA 30 EUR
BLDPBD154575-25G BLDPBD154575-10G BLDPBD154575-500G BLDPBD154575-1G BLDPBD154575-100G BLDPBD154575-5G BLDPBD154575-0.5G
Monomethyl-trans-1,4-cyclohexanedicarboxylate 98%
Monomethyl-trans-1,4-cyclohexanedicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₄O₄
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01311247
CAS-Nummer: 15177-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR