(±)-trans-1,2-Cyclohexandiol 95%

Synonyme: (±)-trans-Cyclohexan-1,2-diol

BD154550-100G BD154550-5G BD154550-10G BD154550-25G
BLDPBD154550-100GEA 138 EUR
BLDPBD154550-100G BLDPBD154550-5G BLDPBD154550-10G BLDPBD154550-25G
(±)-trans-1,2-Cyclohexandiol 95%
(±)-trans-1,2-Cyclohexandiol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₁₂O₂
Molecular Weight: 116,16 g/mol
Lagertemperatur: Ambient
MDL: MFCD00063611
CAS-Nummer: 1460-57-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR