trans,trans-4-Methoxy-4'-propyl-1,1'-bicyclohexyl 97%
|
|
BLDPBD154521-1GEA
13
EUR
BLDPBD154521-1G
trans,trans-4-Methoxy-4'-propyl-1,1'-bicyclohexyl 97%
trans,trans-4-Methoxy-4'-propyl-1,1'-bicyclohexyl
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₆H₃₀O Molecular Weight: 238,42 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12761281 CAS-Nummer: 97398-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...