(trans,trans)-4-Fluorophenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate 97%

Synonyme: rel-4-Fluorophenyl (1r,1's,4R,4'R)-4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate

BD154520-1G BD154520-5G
BLDPBD154520-1GEA 11 EUR
BLDPBD154520-1G BLDPBD154520-5G
(trans,trans)-4-Fluorophenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate 97%
(trans,trans)-4-Fluorophenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₂H₃₁FO₂
Molecular Weight: 346,47 g/mol
Lagertemperatur: Ambient
MDL: MFCD11053422
CAS-Nummer: 81701-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR