trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane) 97%

BD154492-25G BD154492-100G BD154492-5G BD154492-500G
BLDPBD154492-25GEA 30 EUR
BLDPBD154492-25G BLDPBD154492-100G BLDPBD154492-5G BLDPBD154492-500G
trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane) 97%
trans,trans-4-(3,4-Difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane)

Storage condition: Sealed in dry, Room Temperature

Formel: C₂₀H₂₈F₂
Molecular Weight: 306,43 g/mol
Lagertemperatur: Ambient
MDL: MFCD13188635
CAS-Nummer: 118164-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR