meso-Tetraphenylporphin 97%

Synonyme: 5,10,15,20-Tetraphenyl-21H,23H-porphin

BD154434-100G BD154434-10G BD154434-1G BD154434-25G BD154434-250MG BD154434-5G
BLDPBD154434-100GEA 772 EUR
BLDPBD154434-100G BLDPBD154434-10G BLDPBD154434-1G BLDPBD154434-25G BLDPBD154434-250MG BLDPBD154434-5G
meso-Tetraphenylporphin 97%
meso-Tetraphenylporphin

Storage condition: Inert atmosphere, Room Temperature

Formel: C₄₄H₃₀N₄
Molecular Weight: 614,75 g/mol
Lagertemperatur: Ambient
MDL: MFCD00011680
CAS-Nummer: 917-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR