Methyl-N-acetyl-L-tryptophanate 98%

BLDPBD15443-25GEA 117 EUR
BLDPBD15443-25G BLDPBD15443-1G BLDPBD15443-5G BLDPBD15443-100G
Methyl-N-acetyl-L-tryptophanate 98%
Methyl-N-acetyl-L-tryptophanate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₄H₁₆N₂O₃
Molecular Weight: 260,293 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00022759
CAS-Nummer: 2824-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR