7-(Diethylamino)coumarin-3-carboxylic acid 98%

Synonyme: 7-(Diethylamino)-2-oxo-2H-chromene-3-carboxylic acid,DEAC, acid

BD15423-1G BD15423-250MG BD15423-5G BD15423-100G BD15423-25G
BLDPBD15423-1GEA 29 EUR
BLDPBD15423-1G BLDPBD15423-250MG BLDPBD15423-5G BLDPBD15423-100G BLDPBD15423-25G
7-(Diethylamino)coumarin-3-carboxylic acid 98%
7-(Diethylamino)coumarin-3-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₄H₁₅NO₄
Molecular Weight: 261,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD00068721
CAS-Nummer: 50995-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR