Zinn(II)iodid 99.999% (Metall-Basis)

BD154168-1G BD154168-5G BD154168-25G
BLDPBD154168-1GEA 10 EUR
BLDPBD154168-1G BLDPBD154168-5G BLDPBD154168-25G
Zinn(II)iodid 99.999% (Metall-Basis)
Zinn(II)iodid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: I₂Sn
Molecular Weight: 372,52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049545
CAS-Nummer: 10294-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR