Zinn(II)chlorid Dihydrat 99%

Lieferant: BLD PHARMATECH GMBH

Synonyme: Zinnsalz

BLDPBD154165-25GEA 7 EUR
BLDPBD154165-25G
Zinn(II)chlorid Dihydrat 99%
Zinn(II)chlorid Dihydrat

Storage condition: Sealed in dry, Room Temperature

Formel: SnCl₂·2H₂O
Molecular Weight: 225,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149863
CAS-Nummer: 10025-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR