5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine 96%

BD154110-100MG BD154110-250MG BD154110-1G
BLDPBD154110-100MGEA 57 EUR
BLDPBD154110-100MG BLDPBD154110-250MG BLDPBD154110-1G
5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine 96%
5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₄H₁₄ClNS
Molecular Weight: 263,78 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00661081
CAS-Nummer: 55142-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR