Tiagabine hydrochloride 98%

BLDPBD154082-250MGEA 239 EUR
BLDPBD154082-250MG BLDPBD154082-1G BLDPBD154082-100MG
Tiagabine hydrochloride 98%
Tiagabine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₂₀H₂₅NO₂S₂
Molecular Weight: 412,00 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07369025
CAS-Nummer: 145821-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR